Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COQ9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524816
Description
This antibody is predicted to react with rat based on sequence homology.
Coenzyme Q10 (CoQ10), or ubiquinone, is a mobile lipophilic electron carrier critical for electron transfer by the mitochondrial inner membrane respiratory chain. COQ9 is 1 of several enzymes involved in biosynthesis of CoQ10 and likely functions in modification of the benzoquinone ring.
Specifications
COQ9 | |
Polyclonal | |
Unconjugated | |
COQ9 | |
2310005O14Rik; C16orf49; C78387; coenzyme Q9; coenzyme Q9 homolog; coenzyme Q9 homolog (yeast); COQ10D5; COQ9; HSPC326; PSEC0129; Ubiquinone biosynthesis protein COQ9, mitochondrial | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
57017, 67914 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.49 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O75208, Q8K1Z0 | |
COQ9 | |
KLH conjugated synthetic peptide between 156-184 amino acids from the central region of human COQ9. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction