Learn More
Eisco™ Coquina Sedimentary Rock Specimen
SCI_ED
Catalog No. S27607 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27607 12/Pk.
S27585 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27607 Supplier Eisco™ Supplier No. ESNG0078PK12
Add to Cart
Add to Cart
Composed of bone and shell fragments, coquina is nearly 100% fossil matter. Held together by a carbonate cement, this skeletal mass forms in low-energy environments such as lagoons and reefs.
  • Correlates well to Next Generation Science Standards Evidence
  • Great for teaching sedimentary facies, paleontology, and stratigraphy
Specifications
Product Type Sedimentary Rock
Grade K to 16
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.52 kg