Learn More
R&D Systems™ CoraFluor™ 1, thiol reactive
GREENER_CHOICESDP
Catalog No. 811720U Shop All Novus Products
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
81-172-0U 20 μg
1 options
This item is not returnable. View return policy
Catalog No. 81-172-0U Supplier R&D Systems™ Supplier No. 8117/20U
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Terbium cryptate FRET donor for TR-FRET assay development, cysteine reactive for conjugation
CoraFluor™ 1, thiol reactive is a terbium-based time-resolved Förster resonance energy transfer (TR-FRET) donor for TR-FRET assay development, such as target engagement and ternary complex assays. This product is designed with maleimide functionality to conjugate to cysteines, making it suitable for labeling biomolecules, including antibodies, nanobodies, proteins and peptides. It is exceptionally bright, providing unmatched assay sensitivity and stability in biological media over other commercially available TR-FRET donors. CoraFluor™ 1, thiol reactive is compatible with common acceptor dyes such as FAM, TMR and Cyanine 5, SE (Cat. No. 5436), enabling seamless assay integration in high throughput screening applications. CoraFluor™1, amine reactive and CoraFluor™2, amine reactive also available.
Specifications
Quantity 20 μg
Product Line Tocris
Target Fluorescent Probes