Learn More
Honeywell CoreShield Cut Protection Gloves with HPPE/Stainless Steel Lining
Features a 13 Gauge HPPE/Stainless Steel Liner, and a Nitrile Coating. CoreShield Gloves ensure workers are not only protected but also comfortable and productive. A wide variety of additional key features enhance usability, wearability, and durability.
Manufacturer: Honeywell 250913B/8M
Description
- CoreShield High Performance Coating with abrasion enhancement technology doubles glove durability
- Specially designed elastic coating provides outstanding oil- and dry-grip performance
- 360° breathability through an open cell structure provides enhanced comfort, for longer wear
- Prevents hand fatigue for workers who need to wear gloves all day long
- Longer cuff ensures wrist protection
- Reinforced thumb for greater wear resistance and extended product life
- Compatible with resistive touchscreen technology typically used on industrial machines and smartphones
- No significant changes in performance or size even after 5 washing cycles
- Soft, skin-friendly yarns reduce allergy risk for wearers
Specifications
Slip-On | |
13 Gauge | |
HDPE/Stainless Steel | |
Medium | |
Black |
Manufacturing, Automotive, Metal Stamping, Glass Handling, Construction, and Logistics | |
Plastic Bag | |
CoreShield™ | |
Reinforced | |
ANSI 6 |