Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corin Polyclonal Antibody, Abbomax

Catalog No. 502204094
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4094 Supplier Abbomax Inc Supplier No. 602650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Mouse heart tissue Cellular location: Cell Membrane.

Corin Polyclonal antibody specifically detects Corin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Corin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene CORIN
Gene Accession No. Q80YN4, Q9Y5Q5, Q9Z319
Gene Alias ATC2; Atrial natriuretic peptide-converting enzyme; Atrial natriuretic peptide-converting enzyme, 100 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, 160 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, 180 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, activated protease fragment; Atrial natriuretic peptide-converting enzyme, N-terminal propeptide; AV273130; Corin; corin, serine peptidase; CRN; EC 3.4.21; Heart-specific serine proteinase ATC2; low density lipoprotein receptor-related protein 4; low density lipoprotein-related protein 4; Lrp4; MGC119742; PEE5; pro-ANP-convertase; pro-ANP-converting enzyme; PRSC; serine peptidase; serine protease; TMPRSS10; Transmembrane protease serine 10
Host Species Rabbit
Immunogen A synthetic peptide sequence derived from 300aa-450aa of mouse corin protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10699, 289596, 53419
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.