Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CORIN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107104 Supplier Invitrogen™ Supplier No. PA5107104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 93%; Zebrafish: 100%.

CORIN is a member of the type II transmembrane serine protease class of the trypsin superfamily. Members of this family are composed of multiple structurally distinct domains. CORIN converts pro-atrial natriuretic peptide to biologically active atrial natriuretic peptide, a cardiac hormone that regulates blood volume and pressure. This protein may also function as a pro-brain-type natriuretic peptide convertase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CORIN
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene CORIN
Gene Accession No. Q9Z319
Gene Alias ATC2; Atrial natriuretic peptide-converting enzyme; Atrial natriuretic peptide-converting enzyme, 100 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, 160 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, 180 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, activated protease fragment; Atrial natriuretic peptide-converting enzyme, N-terminal propeptide; AV273130; Corin; corin, serine peptidase; CRN; EC 3.4.21; Heart-specific serine proteinase ATC2; low density lipoprotein receptor-related protein 4; low density lipoprotein-related protein 4; Lrp4; MGC119742; PEE5; pro-ANP-convertase; pro-ANP-converting enzyme; PRSC; serine peptidase; serine protease; TMPRSS10; Transmembrane protease serine 10
Gene Symbols CORIN
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Mouse Corin.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53419
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.