Learn More
R&D Systems™ CORM 3
GREENER_CHOICESDP
Catalog No. 532050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-205-0 50 mg
53-201-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-205-0 Supplier R&D Systems™ Supplier No. 5320/50
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Water-soluble carbon monoxide-releasing molecule
CORM 3 is a water-soluble carbon monoxide-releasing molecule. Suppresses thrombin-induced nitrite release in BV-2 microglia. Protects isolated rats hearts from reperfusion damage in vitro and is cardioprotective in a rat myocardial infarction model in vivo. Also prolongs survival of transplanted hearts in mice. Exhibits anti-inflammatory and vasorelaxant effects.
Specifications
Quantity 50 mg
Product Line Tocris
Product Type CORM 3
Molecular Weight (g/mol) 294.61