Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corn oil

Catalog No. p-3598342
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405435000 500 g Glass bottle
AC405430025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC405435000 Supplier Thermo Scientific Chemicals Supplier No. 405435000
Only null left
Add to Cart

CAS: 8001-30-7

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-30-7
MDL Number MFCD00130865

Specifications

Density 0.9200g/mL
Flash Point >200°C
Packaging Glass bottle
Refractive Index 1.4730 to 1.4760
Quantity 500 g
Specific Gravity 0.92
Merck Index 14, 2533
Solubility Information Solubility in water: insoluble
Chemical Name or Material Corn oil
EINECSNumber 232-281-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.