Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corn Oil, NF, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
25 mL
500 mL
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Description
Spectrum™ Chemical Corn Oil, NF its uses include as a solvent vehicle for topical preparations and as a glidant excipient for oral dosage tablets and capsules. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 100% |
Packaging | Drum |
Quantity | 200 L |
Grade | NF |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction