Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ 384-Well Nonbinding Surface (NBS™) Microplates
Shop All Corning Products

Click to view available options
Color:
Black
White
Well Color:
Black
White
Barcode:
Includes Barcode
Description
- Ideal for homogenous assays such as Scintillation Proximity Assays (SPA) and fluorescent assays
- Modified polymer surface is nonionic hydrophilic, minimizing bio-interaction within wells to reduce nonspecific binding and reagent costs
- Significantly reduces protein and nucleic acid binding to polymers, maintains enzyme activity, and inhibits adhesion of a number of cell lines Stable to aqueous solutions containing EtOH
- Use at temperature range of 4° to 37°C (39° to 99°F)
- Flat-bottom wells
- Compatible with a wide range of plate readers, liquid handlers, and robotic systems
Generic Bar Coded Plates
- Code 128 bar code format ensures compatibility with most bar code scanning and software systems
- To avoid mix-ups, the surface treatment of the microplate wells is easily identified
- Labels applied to all four sides of the plate No sequence number duplicates
Specifications
Specifications
| Barcode | Includes Barcode |
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Color | White |
| For Use With (Equipment) | Plate readers, liquid handlers and robotic systems |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 20 to 80 μL |
| Sterility | Non-sterile |
| Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction