Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ BioCoat™ #1 German Glass Coverslips, Round
Shop All Corning Products

Click to view available options
Diameter (Metric):
12 mm
22 mm
Surface Treatment:
Collagen Type I
Fibronectin
Laminin/Poly-D Lysine
Poly-D Lysine
Poly-L Lysine
Description
Corning™ BioCoat™ German Glass coverslips with surface treatment (see individual Specification tables for coating). Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
Shelf life: ≥ 3 months from date of shipment

Specifications
Specifications
Type | Slide Coverslip |
Surface Treatment | Poly-D Lysine |
Material | No. 1 German Glass |
Diameter (English) | 0.472 in. |
For Use With (Application) | Fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy, phase contrast microscopy |
Diameter (Metric) | 12 mm |
No. per Case | 80 |
No. per Pack | 80 |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction