Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap
Shop All Corning Products

Click to view available options
Capacity (Metric):
70 mL
250 mL
600 mL
750 mL
Culture Area:
150 cm2
175 cm2
25 cm2
75 cm2
No. per Case:
10/Cs.
40/Cs.
5/Cs.
50/Cs.
Neck Style:
Canted
Straight
Description
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
Applications include:
- Promotion of cell attachment and spreading
- Rapid expansion of cell populations
- Serum-free or reduced serum culture
- Cell adhesion assays
- Improving survival of primary cells in culture
Source
Quality Control
- Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
- Tested and found negative for bacteria and fungi
- Collagen I purity >90% by SDS-PAGE
Storage
Specifications
Specifications
| Barcode | Yes |
| Capacity (English) | 20.2 oz. |
| Closure Material | Phenolic |
| Capacity (Metric) | 600 mL |
| For Use With (Application) | For adhesion, growth and differentiation of neuronal and transfected cell lines |
| Neck Style | Canted |
| No. per Case | 5/Cs. |
| No. per Pack | 5/Pk. |
| Shape | Rectangular |
| Surface Coating | Collagen I |
| Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction