Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ BioCoat™ Poly-L-Lysine Culture Dishes
Shop All Corning Products
Click to view available options
Diameter (Metric):
35 mm
60 mm
No. per Pack:
20/pk.
5/pk
No. per Case:
100/cs.
20/cs.
Description
Suitable for promoting adhesion of transfected cell lines and other fastidious cell types. Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology.
- Minimize assay failure due to cell detachment
- Matrix components are applied as an even, optically clear coating
- Precoated with Poly-L-Lysine to eliminate time-consuming preparation
- Lot-to-lot consistency provides reproducible experiments
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For adhesion, growth and differentiation of neuronal and transfected cell lines
Order Info
Bulk quantities available; please contact your Customer Service Representative.

Specifications
Specifications
Bottom Shape | Round |
Culture Area | 9.6 sq. cm |
Shape | Round |
Surface Treatment | Poly-L-Lysine |
Type | Cell Culture Dish |
No. per Case | 20/cs. |
No. per Pack | 20/pk. |
Diameter (Metric) | 35 mm |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction