Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ 96-Well, Poly-L Lysine-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 0877292
Click to view available options
Packaging:
5/Cs
50/Cs

Color: Clear; Sterility: Sterile; Lid: With Lid

Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.

Applications include:

  • Attachment and spreading of a variety of cell types
  • Cell differentiation and neurite outgrowth
  • Attachment of fastidious transfected cell lines
  • Support survival of primary neurons in culture
  • Serum-free or reduced-serum culture

Source

  • PDL, synthetic (MW 75-150 kD)
  • PLL, synthetic (MW 30-70 kD)

Quality Control

  • Tested for ability to promote firm attachment of RCG cells
  • Tested and found negative for bacteria and fungi

Storage

  • 4°C to 30°C under dry conditions.
  • Order Info

    Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates

    TRUSTED_SUSTAINABILITY

    Specifications

    Format Standard
    Lid With Lid
    Content And Storage 4°C to 30°C
    Culture Area 0.32 cm2
    Array 8 x 12
    Packaging 5/Cs
    Color Clear
    Material Polystyrene
    No. of Wells 96
    Sterility Sterile
    Surface Treatment Poly-L-Lysine
    Well Shape Round, Flat Bottom (F-well)
    For Use With (Application) Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
    Quantity 5/Cs.
    Source Poly-L-Lysine
    Well Color Clear
    Show More Show Less
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.