Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Black and White 96-Well Polypropylene Assay Plates
Shop All Corning Products

Click to view available options
Color:
Black
White
Description
- Ideal for fluorescence and luminescence assays and for compound storage
- Round-bottom wells
- Untreated surface

Specifications
Specifications
Format | Standard |
Lid | Without Lid |
Array | 8 x 12 |
Color | Black |
Material | Polypropylene |
No. of Wells | 96 |
Volume (Metric) Working | 75 to 200 μL |
Sterility | Non-sterile |
Surface Treatment | Non-treated |
Well Shape | Round Bottom (U-well) |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction