Learn More
Corning™ Bovine Pituitary Extract (BPE)
GREENER_CHOICE
Catalog No. p-90810
Click to view available options
Quantity:
15 mg
Supplement used in the culture of a variety of epithelial and endothelial cells
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2° to 8°C
Product Type Bovine Pituitary Extract
Quantity 15 mg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.