Learn More
Corning™ Endothelial Cell Growth Supplement (ECGS)

Shop All Corning Products


Description
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
- Vialed, highly purified growth factors with high biological activity
- Broad range of highly purified natural and recombinant lymphokines
- Enriched culture supplements to be reconstituted/added in the medium of your choice
- Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2° to 8°C |
| Description | ECGS is a broadly used supplement to culture a variety of cells, particularly endothelial cells. ECGS contains various growth factors (e.g., acidic FGF or ECGF-a) |
| Concentration | 50 to 150 µg/ mL depending on cell type. May require supplementation of 1 to 100 µg/mL heparin. |
| Product Type | Growth Factor |
| Formulation | Lyophilized from NaCl-containing buffer with streptomycin sulfate; reconstitute in serum-free medium |
| Quantity | 100 mg |
| Source | Bovine brain |
| Tested For | Tested for ability to promote proliferation of Fetal Bovine Heart Endothelial Cells (FBHEC) in medium containing 10% newborn calf serum. Filtered (0.2 µm membrane) and tested and found negative for bacteria, fungi and mycoplasma. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.