Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Gentamicin Sulfate
GSA_VA

Catalog No. p-4877179 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 x 100 mg
1 x 1 g
10 x 10 mL
Form:
Liquid
Powder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MT30005CR 10 x 10 mL Liquid
MT61098RA 1 x 100 mg Powder
MT61098RF 1 x 1 g Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MT30005CR Supplier Corning™ Supplier No. 30005CR
Only null left
Add to Cart
Edge
Add to Cart

Packaged in plastic bottles

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Concentration 50 mg/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile
Format Plastic Bottle
Quantity 10 x 10 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.