Learn More
Corning™ Ham's Medium
GSA_VA
Catalog No. p-2410154
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
Product Type:
Ham's F-10
Ham's F-12 Medium (Mod.) 1 X
This item is not returnable. View return policy
This item is not returnable. View return policy
Sterile, with L-Glutamine
Lower documented Endotoxin content than offerings from other major suppliers. Ham's F-12 media was originally formulated for the serum-free culture of CHO cells, and is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids.  This medium is also suitable for carcinoma cells, rat skeletal myoblasts, Chinese hamster lung cells, and rat, rabbit, and chicken embryos.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Ham's F-10
Sterility Sterile
With Additives Glutamine
Quantity 6 x 500 mL
pH 7 to 7.4
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.