Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Iscove's Modification of DMEM

Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
With Additives:
Glutamine, HEPES
HEPES
Without Additives:
No α-Thioglycerol, No beta-Mercaptoethanol
No Glutamine, No beta-Mercaptoethanol
Description
Substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over the original formulation. Iscove's Modification of DMEM (IMDM) is a highly enriched synthetic medium optimized for rapidly proliferation cultures of high cell density. A modified form of Dulbecco's Modified eagle Medium (DMEM), Iscove's contains additional amino acids, vitamins, selenium and HEPES buffer, and is formulated with potassium nitrate in place of ferric nitrate. The addition of albumin and transferrin may be required for certain cell types. This formulation is produced with L-glutamine and 25 mM HEPES, and do not contain alpha-thioglycerol and beta-mercaptoethanol.

Specifications
Specifications
Concentration | 1 X |
Form | Liquid |
Product Type | Iscove's Modification of DMEM media |
Sterility | Sterile |
With Additives | Glutamine, HEPES |
Without Additives | No α-Thioglycerol, No beta-Mercaptoethanol |
Quantity | 6 x 1 L |
pH | 7 to 7.4 |
Content And Storage | 2° to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction