Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Clear Polypropylene 384-Well Deep-Well Plates

Click to view available options
Sterility:
Non-sterile
Sterile
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
Order Info
Packaged in bags of ten
Certifications
Certified DNase- and RNase-free

Specifications
Specifications
Format | Standard |
Lid | Without Lid |
Array | 16 x 24 |
Packaging | Bags of 5 |
Color | Clear |
Product Type | 384 Deep-Well Microplate |
Material | Polypropylene |
Volume (Metric) Well | 240 μL |
No. of Wells | 384 |
Sterility | Non-sterile |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction