Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Collagen I, Human
GREENER_CHOICE

Catalog No. p-4696753 Shop All Corning Products
Click to view available options
Quantity:
10 mg

Catalog No. 08-774-550

Supplier: Corning™ 354265

Only null left
Add to Cart
Add to Cart

Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.2 to 2 μg/cm2
For Use With (Application) Cell Attachment and Proliferation, Cell-specific Morphology and Function
Formulation Frozen in 2 mm hydrogen chloride
Quantity 10 mg
Source Human Placenta
Product Type Collagen, Human Type I
Content And Storage -20°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.