Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Cover Glasses

Catalog No. p-45435 Shop All Corning Products
Click to view available options
Dimensions (L x W):
30 x 24 mm
Shape:
Rectangle

Fine optical glass developed specifically for cover glasses

  • Free of stones, blisters, bubbles and striae
  • High resistance to surface attack, weather

Order Info

  • Packaged in lint-free reclosable plastic boxes for complete protection.
  • Supplied in packs of 1 oz.

TRUSTED_SUSTAINABILITY

Specifications

Shape Rectangle
Dimensions (L x W) 30 x 24 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.