Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Untreated Culture Dishes
BIOPHARMAGSA_VAGREENER_CHOICE

Catalog No. 0877232 Shop All Corning Products
Click to view available options
Diameter (Metric):
35 mm
60 mm
100 mm
150 mm
Culture Area:
148 cm2
21 cm2
55 cm2
8 cm2
Height (Metric):
10 mm
15 mm
20 mm
25 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Binds bio-molecules through passive interactions and hydrophobic in nature. Corning™ Untreated Culture Dishes provide an ideal environment for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.

  • Manufactured from optically clear virgin polystyrene
  • Not treated for applications where cell attachment is not desired
  • Not treated for applications where cell attachment is not desired
  • Stacking beads aid in handling
  • Vents provide consistent gas exchange
  • Sterile
  • Sterile
  • Nonpyrogenic


Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.

Cat. no. 431111, Square dishes with interior bottom dimensions of 224 x 224 mm.

Order Info

For Cell Culture Treated Polystyrene Dishes: Shelf pack 430293 consists of six-pack carriers which hold six sleeves of 10 dishes each–minimizing contamination of unused dishes after sleeve is opened. Supplied in sleeves of 20 dishes (except 150 x 25mm dish which is packaged five per sleeve and 245 x 245mm dish which comes four per sleeve).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 16.5 mL (Well)
Color Clear
Culture Area 55 cm2
Disposable Single-use
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Non-treated
Type Cell Culture Petri Dish
Includes Lid
No. per Case 500/Cs.
No. per Pack 20/Pk.
Height (Metric) 20 mm
Diameter (Metric) 100 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.