Learn More
Corning™ DeckWorks™ Low Binding Hinged Rack Barrier Tips
Shop All Corning Products

Description
Corning DeckWorks low binding barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Volume (Metric) | 0.1 to 10 μL |
Filtered | Filtered |
Sterility | Sterile |
Color | Natural |
For Use With (Application) | Used for quantitative assays in which sample binding can significantly alter results |
Format | Hinged Rack |
Tip Style | Low Retention, Round |
Packaging | Rack |
Graduated | Yes |
Product Line | DeckWorks |
Show More |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.