Learn More
Corning™ Digital Display and Stirrer Accessory Kit
Shop All Corning Products
Click to view available options
Type:
Hot Plate Kit
Description
Includes
Temperature controller (6795PR) that controls the temperature of liquids up to 200°C
- Voltage independent
- Two piece, 0.31 in. (0.8cm) diameter, 18 in. (45.7cm) long support rod (440129) and magnetic stir bar retriever (6970SR)
Specifications
Specifications
For Use With (Equipment) | PC-400D, PC-420D, PC-600D, PC-620D Digital Display Hotplates and Stirring Hotplates |
Includes | Temperature Controller (6795PR), A Two Piece, 0.8 cm Diameter, 45.7 cm L Support Rod (440129) and Magnetic Stir Bar Retriever (6970SR) |
Type | Hot Plate Kit |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov