Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Disposable Glass Pasteur Pipets, Non-Sterile, Unplugged
GSA_VA

Catalog No. p-3207917 Shop All Corning Products
Click to view available options
Length (Metric):
14.6 cm
22.86 cm
33.02 cm
Diameter (Metric) Outer:
0.55 cm
0.65 cm
This item is not returnable. View return policy

Catalog No. 13-678-6

Supplier: Corning™ 7095BNMR

Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.

Corning disposable glass Pasteur pipets are available in convenient Packages. One-time use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents. These pipets feature a constriction one inch below the top to facilitate plugging. They are ideal for rapid non­volumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.



'B' series pipets are soda lime glass. 'D' series are borosilicate glass.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Aspirator Plug No
Diameter (English) Outer 0.255 in.
Diameter (Metric) Outer 0.65 cm
Material Soda Lime Glass
Includes 1 in. Constriction below the top
Length (English) 13 in.
Tip Style Round
Length (Metric) 33.02 cm
Type Unplugged Pasteur Pipet
Disposable Single-use
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.