Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ DNA-BIND™ 96-Well Plates

Catalog No. p-154112 Shop All Corning Products
Click to view available options
Color:
Clear
Quantity:
50/Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Convenient method for covalently immobilizing ssDNA by either the 5' or 3' end

  • Ready to use without surface preparation, prehybridization, or addition of expensive crosslinking reagents or highly purified oligos
  • Ideal for hybridization and amplification
  • Surface provides binding with high specificity and extremely low background
  • Flat-bottom wells

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) Ideal for hybridization and amplification
No. per Case 50/Cs.
No. per Pack 1/Pk.
Quantity 50/Cs.
Well Color Clear
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.