Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Epidermal Growth Factor (EGF), Mouse Natural (Receptor Grade)
GSA_VAGREENER_CHOICE

Catalog No. p-96954 Shop All Corning Products
Click to view available options
Quantity:
100 μg
Purity or Quality Grade:
>95% by SDS-PAGE and >80% by pH 9.9 PAGE
>98% by SDS-PAGE and pH 9.9 PAGE

Catalog No. CB-40001

Supplier: Corning™ 354001

Only null left
Add to Cart
Add to Cart

Enhance the proliferation of a wide variety of cell types

The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.

  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions

Growth factors are proteins that bind to receptors found on the surface of non-hematopoietic cells. Each family of growth factors affect specific cell types, e.g., nerve growth factors (NGF) affect nerve cell types, and epidermal growth factors (EGF) affect epithelial cell types.

Corning offers a range of growth factors that are derived from different species, both natural and recombinant. These growth factors have been used to promote long-term proliferation in cell culture and to induce cell differentiation in a variety of cell types under serum-free or reduced serum conditions.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Product Type Growth Factor
Format Natural
Formulation Lyophilized from ammonium acetate, carrier free, reconstitute in dH2O
For Use With (Application) A broad range of highly purified natural and recombinant lymphokine and growth factor biochemicals designed specifically for use in basic cell biology research
Purity or Quality Grade >95% by SDS-PAGE and >80% by pH 9.9 PAGE
Quantity 100 μg
Reactivity Mouse
Source Male Mouse Submaxillary Glands
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.