Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CellBIND™ HYPERStack™ Culture Vessels
BIOPHARMA

Catalog No. p-4186328 Shop All Corning Products
Click to view available options
Culture Area:
6000 cm²
Product Type:
12-layer vessel
Quantity:
4/Cs.

Catalog No. 07-201-843

Supplier: Corning™ 10012

Closed system that allows fluid manipulations to occur through preassembled tubing sets

Closed System for High Yield Cell Growth

Introducing the next generation in Corning’s High Yield PERformance (HYPER) platform – the Corning HYPERStack cell culture vessel. This product line combines the best of two Corning products: the Corning CellSTACK™ culture chamber and the Corning HYPERFlask™ vessel. The utilization of the proprietary gas-permeable film technology provided in the format of the CellSTACK culture chamber allows the HYPERStack vessel to be the most efficient, scalable cell culture vessel for adherent cell culture available today.

Features and Benefits

  • More cells – 2.5x more cells per volumetric footprint
  • Closed system – no open fluid manipulations
  • Scalable product – multiple size offerings
  • Ergonomic design – easier manipulation
  • Fixed media volume – 0.2 mL/cm² fills vessel for optimal handling
  • Innovative assembly – no adhesives, low particulate
  • Less volumetric waste – less waste per growth area lowers overall costs

Surface Treatment Guide

The Corning CellBIND surface is designed to enhance cell attachment and increase cell yields. This enhanced surface treatment can often improve cell attachment of loosely adherent cells and maintain growth in reduced-serum or serum-free conditions. Alternatively, a Non-CellBIND option (named as Untreated) can support conventional cell attachment and allow for a faster and gentler harvest procedure (i.e., growing and harvesting cells for downstream assays).

For more information, please refer to the Corning™ CellBIND™ Surface Brochure in the resource tab on the product pages below.

Protocol Guides

Select HYPERStack video protocols available at www.corning.com/HYPER

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Material Polycarbonate
No. per Case 4/Cs.
No. per Pack 1/Pk.
Product Type 12-layer vessel
Graduated No
Sterility Sterile
Bottom Shape Rectangular
Culture Area 6000 cm²
Quantity 4/Cs.
Sterilization Method Gamma sterilized to SAL 10-6 standard
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.