Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ PureCoat™ Amine Culture Dishes

Click to view available options
No. per Case:
10/Cs.
40/Cs.
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.

Specifications
Specifications
Bottom Shape | Flat |
Culture Area | 58.1 cm2 |
Material | Polystyrene |
Shape | Round |
Surface Treatment | Amine |
Type | Cell Culture Dish |
No. per Case | 10/Cs. |
No. per Pack | 10/Pk. |
Diameter (Metric) | 100 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction