Learn More
Corning™ PureCoat™ Carboxyl Multiwell Plates
GREENER_CHOICE
Catalog No. 08772166 Shop All Corning Products
Change view
Click to view available options
Well Count:
6
Catalog No. Well Count
08-772-166 6
1 options
Catalog No. 08-772-166 Supplier Corning™ Supplier No. 354773
Only null left
Add to Cart
Add to Cart
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
TRUSTED_SUSTAINABILITY
Specifications
Array 2 x 3
Certifications/Compliance ANSI, SBS
Bottom Shape Flat
Material Polystyrene
Shape Rectangular
Sterility Sterile
Type Multi-well Cell Culture Plate
Well Count 6
For Use With (Application) Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested
No. per Case 5/Cs.
No. per Pack 5/Pk.
Quantity 5/Cs.
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov