Learn More
Corning™ PureCoat™ Collagen I Peptide TC-Treated Microplates
Shop All Corning Products

Description
Includes
Lid
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
Specifications
Specifications
| Certifications/Compliance | cGMP |
| Closure Color | Clear |
| Closure Included | Yes |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Color | Clear |
| Culture Area | 2 cm2 |
| Material | Polystyrene |
| Shape | Rectangular |
| Sterility | Sterile |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.