Learn More
Corning™ PureCoat™ Collagen Peptide Cell Culture Flasks with Vented Cap
GREENER_CHOICE
Catalog No. 08774356
Click to view available options
Culture Area:
175 cm2
75 cm2
Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
Neck Style Canted
No. per Case 10/Cs.
No. per Pack 5/Pk.
Shape Rectangular
Surface Coating PureCoat E cm Mimetic Collagen I Peptide
Bottom Shape Rectangular
Closure Type Vented
Culture Area 75 cm2
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.