Learn More
Corning™ PureCoat™ E cm Mimetic Multi-Flask
GREENER_CHOICE
Catalog No. p-4819760 Shop All Corning Products
Change view
Click to view available options
Surface Treatment:
Collagen I Peptide
Fibronectin Peptide
Culture Area:
525 cm2
875 cm2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment Culture Area
07-201-377 Fibronectin Peptide 525 cm2
07-201-378 Fibronectin Peptide 875 cm2
07-201-379 Collagen I Peptide 525 cm2
07-201-380 Collagen I Peptide 875 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-201-377 Supplier Corning™ Supplier No. 356244
Only null left
Add to Cart
Add to Cart
Features multi-layers with vented cap
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
Compliance
cGMP manufacturing compliant (ISO 9001:2008, ISO 13485:2012)
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Closure Material Polypropylene
Description Individually packed in foil bag
Neck Style Straight
No. per Case 8/Cs.
No. per Pack 1/Pk.
Shape Rectangular
Surface Coating PureCoat E cm Mimetic Fibronectin Peptid
Sterility Sterile
Bottom Shape Rectangular
Closure Type Vented
Culture Area 525 cm2
Surface Treatment Fibronectin Peptide