Learn More
Corning™ PYREX™ VISTA™ Class A Cylinder
SCI_ED
Catalog No. p-3004053 Shop All Corning Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Calibrated To Contain.
  • Blue enamel graduations
  • Capacity is in accordance with ASTM™ E-1272
  • 10mL size has a funnel top for easier filling
TRUSTED_SUSTAINABILITY
Specifications
Sterility Non-sterile
Certifications/Compliance ASTM E1272
Type Graduated Cylinder
Class A
Scale Color Blue
Calibration To Contain
Material Borosilicate Glass
Capacity (Metric) 25 mL
Includes Funnel Top, Pourout
Scale Single
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.