Description
- Large, blue, block letters make inscriptions easy to read
- Sharp, permanent graduation lines
- Snap caps are supplied with all sizes
Specifications
Specifications
| Calibration | Calibrated To Contain |
| Closure Color | Blue |
| Closure Material | LDPE |
| Capacity (Metric) | 25 mL |
| Material | Glass, Borosilicate Glass |
| Neck Style | Long |
| Bottom Shape | Flat |