Description
- Full length standard taper outer joints
- Short necks
- Manufactured from No. 7740 borosilicate glass
- Wall thicknesses provide balance between thermal expansion and mechanical strength
Compliance
Designed to ASTM™ E-1403.
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass |
| Neck Style | Short |
| Standard Taper Joint | 24/40 |
| Disposable | Reusable |
| Bottom Shape | Round |