Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Single-Metric Scale Reusable Graduated Cylinders Available on GSA/VA Contract for Federal Government customers only.

Transluscent polypropylene with raised graduations at primary scale points

Manufacturer:  Corning™ 3022P2L

 View more versions of this product

Catalog No. 10-210-657


  Check Availability
Add to cart

Specifications

Description

  • Hexagonal base
  • Calibrated “To Contain” (TC)
  • Tolerances according to ISO 6706

Exposure to temperatures above 60°C may affect accuracy.

Description & Specifications

Specifications

2000mL
±20mL
Hexagonal
No
18.97 in.
482mm
Yes
20mL
PP (Polypropylene)
ISO 6706
To contain
3.3 in.
8.4cm
20mL
60°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit