Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Square Polycarbonate Storage Bottles with 45 mm Cap
Shop All Corning Products

Click to view available options
Capacity (Metric):
150 mL
250 mL
500 mL
1 L
Description
Includes
Linerless, one-piece, autoclavable orange GL45 threaded polypropylene plug-seal caps
Corning™ square polycarbonate storage bottles are easier to handle, require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing, sampling and storage.
Caution: These square polycarbonate bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.
Strong polycarbonate bottles are more break resistant than other glass or plastic bottles.
Use a linerless, 45mm one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.
Screened white enamel graduations are easier to see than molded graduations
Sterilized by gamma radiation
Large white marking spot for easier identification
Bottles can be autoclaved once at 121oC and 15psi. Repeated autoclaving weakens polycarbonate and is not recommended.
Optional reusable caps with silicone septa (Corning Cat. Nos. 1395-45HTSC, 1395-45SS, 1395-45TS) for syringe sampling or introduction of reagents are available for these caps.
Note: Most GL45 threaded caps designed for glass storage bottles use a different sealing design and will not give a secure seal if used on these plastic bottles unless they have a rubber liner.
Caution: These square polycarbonate bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.

Specifications
Specifications
Capacity | 30 oz., 1000 mL |
Capacity (Metric) | 1 L |
Capacity (English) | 30 oz. |
Material | Polycarbonate |
No. per Case | 24/Cs. |
No. per Pack | 12/Pk. |
Sterility | Sterile |
Closure Color | Orange |
Graduations | 50 to 150 mL |
Autoclavable | Autoclavable |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction