Learn More
Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates
Shop All Corning Products

Description
- Low protein-binding surface prevents loss of sample
- For oil-free operation, use polyethylene or aluminum sealing tape in conjunction with heated-lid thermal cyclers; also accommodates silicone reusable sealing mat
- Withstands temperatures to 135°C without warping
- Various well shapes fit popular thermal cyclers
- Surface of DNA-BIND™ plate allows covalent attachment of ssDNA at 5« and 3« end
- Nonsterile
Certifications
Polypropylene plates are certified DNase- and RNase-free.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Color | Clear |
| Product Type | PCR Microplate, Model P |
| For Use With (Equipment) | Applied Biosystems GeneAmp™ PCR System 9600b, Barnstead Thermolyne Amplitron II™, Techne™ Cyclogene, and Gene E with 96 x 0.2 mL block |
| Material | Polycarbonate |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Well Shape | Round |
| Show More |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.