Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Universal Fit Hinged Rack Pipet Tips, Nonsterile
GREENER_CHOICE

Catalog No. p-3142362 Shop All Corning Products
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Color:
Blue
Yellow
Format:
Hinged Rack
Rack Insert

Hinged closure provides easy stacking and steam penetration during autoclaving

  • Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
  • Beveled orifice helps ensure accurate fluid delivery.
  • 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
  • Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
  • RNase-/DNase-free.
  • Nonpyrogenic.

Order Info

Available sterile and nonsterile

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 to 1000 μL
Sterility Non-sterile
Color Yellow
For Use With (Application) Hinged closure provides easy stacking and steam penetration during autoclaving
Format Hinged Rack
Tip Style Round
Packaging Hinged rack
Graduated Yes
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.