Learn More
CORO1A Mouse anti-Human, Clone: OTI1A5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501429

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Alternative splicing results in multiple transcript variants. A related pseudogene has been defined on chromosome 16.Specifications
CORO1A | |
Monoclonal | |
Unconjugated | |
CORO1A | |
CLABP, CLIPINA, HCORO1, IMD8, TACO, p57 | |
Mouse | |
Affinity Chromatography | |
RUO | |
11151 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1A5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P31146 | |
CORO1A | |
Full length human recombit protein of human CORO1A produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.