Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CORO1B Antibody (OTI3D10), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272086FR
Description
CORO1B Monoclonal specifically detects CORO1B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CORO1B | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
CORO1B | |
Full length recombinant protein of human CORO1B (NP_065174) produced in HEK293T cells. | |
0.1 mL | |
57175.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI3D10 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
coronin, actin binding protein, 1B, coronin, actin-binding protein, 1B, coronin-1B, coronin-2, DKFZp762I166 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction