Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CORO1B Mouse anti-Human, Clone: OTI3D10, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA501573S

Description
Members of the coronin family, such as CORO1B, are WD repeat-containing actin-binding proteins that regulate cell motility.Specifications
CORO1B | |
Monoclonal | |
0.64 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9BR76 | |
CORO1B | |
Full length human recombit protein of human CORO1B produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI3D10 | |
Unconjugated | |
CORO1B | |
CORONIN-2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
57175 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction