Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CORO2A Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299188JF549
Description
CORO2A Polyclonal specifically detects CORO2A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CORO2A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CLIPINB, coronin, actin binding protein, 2A, coronin, actin-binding protein, 2A, coronin-like protein B, DKFZp686G19226, IR10coronin 2A, WD protein IR10, WD repeat-containing protein 2, WDR2coronin-2A, WD-repeat protein 2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human CORO2A. | |
0.1 mL | |
Signal Transduction | |
7464 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction