Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CORO2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215967
Description
CORO2A Polyclonal specifically detects CORO2A in Human samples. It is validated for Western Blot.Specifications
CORO2A | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
CLIPINB, coronin, actin binding protein, 2A, coronin, actin-binding protein, 2A, coronin-like protein B, DKFZp686G19226, IR10coronin 2A, WD protein IR10, WD repeat-containing protein 2, WDR2coronin-2A, WD-repeat protein 2 | |
Rabbit | |
60 kDa | |
0.1 mL | |
Signal Transduction | |
7464 | |
Human | |
IgG |
Western Blot | |
0.99mg/mL | |
Western Blot 1:500-1:3000 | |
Q92828 | |
CORO2A | |
Recombinant protein encompassing a sequence within the center region of human Coronin 2A. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Bovine (82%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction