Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CORO7 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318451UV
Description
CORO7 Polyclonal antibody specifically detects CORO7 in Human samples. It is validated for Western BlotSpecifications
CORO7 | |
Polyclonal | |
Western Blot | |
coronin-7, 0610011B16Rik, coronin 7, CRN7, POD1 | |
Recognizes region between residues Between Between 875 and 925.. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
79585 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction