Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CORO7-PAM16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114238
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114238 Supplier Invitrogen™ Supplier No. PA5114238
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NYEHLFKVND KSVGGSFYLQ SKVVRAKERL DEELKIQAQE DREKGQMPHT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 86%; Human: 100%; Mouse: 86%; Pig: 79%; Rat: 93%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CORO7-PAM16
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CORO7
Gene Accession No. P57737, Q9Y3D7
Gene Alias 0610011B16Rik; 70 kDa WD repeat tumor rejection antigen homolog; CGI-136; CORO7; coronin 7; coronin-7; Crn7; MAGMAS; magmas-like protein; mitochondria associated protein involved in granulocyte macrophage colony stimulating factor signal transduction; mitochondria-associated granulocyte macrophage CSF-signaling molecule; Mitochondrial import inner membrane translocase subunit TIM16; PAM16; POD1; presequence translocase-associated motor 16 homolog (S. cerevisiae); Presequence translocated-associated motor subunit PAM16; SMDMDM; TIM16; TIMM16
Gene Symbols CORO7, PAM16
Host Species Rabbit
Immunogen Immunogen sequence: NYEHLFKVND KSVGGSFYLQ SKVVRAKERL DEELKIQAQE DREKGQMPHT.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51025, 79585
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.