Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coronavirus Antibody (FIPV3-70) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$206.50 - $501.50
Specifications
Antigen | Coronavirus |
---|---|
Clone | FIPV3-70 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), ELISA, Immunofluorescence, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017635
![]() |
Novus Biologicals
NB100647540.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10064754
![]() |
Novus Biologicals
NB10064754 |
0.125 mg |
Each for $501.50
|
|
|||||
Description
Coronavirus Monoclonal specifically detects Coronavirus in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Coronavirus | |
1.0 mg/mL | |
Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), ELISA, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Virus | |
IgG2a | |
Protein A purified | |
NB100-64754 reacts with feline infectious peritonitis virus (FIPV) type 1 and 2 and is known to be specific for the nucleocapsid. It is also known to react with canine coronavirus (CCV), pig coronavirus transmissible gastroenteritis virus (TGEV) and ferret coronavirus. Some specific activity has been detected against bovine coronavirus (BCV). Clone FIPV3-70 exhibits negative reactivity against Feline Leukemia virus, Feline Immunodeficiency virus, Feline Calcivirus, Feline Herpes virus, Canine Adenovirus (type 2), Canine Distemper virus, Canine Parvovirus and Canine Parainfluenza virus. |
FIPV3-70 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Coronavirus cocktail | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title